Order Entry
Germany
ContactUsLinkComponent
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide ≥98%, Millipore®
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide ≥98%, Millipore®
Catalog # 454454-50
CAS Number:  
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide ≥98%, Millipore®
Catalog # 454454-50
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥98% by HPLC
  • Form:
    Off-white solid
  • Solubility:
    Soluble in DMF (5 mg/ml), DMSO (5 mg/ml), and ethanol (1 mg/ml)
  • Storage:
    +2 to –8 °C
  • Stability:
    Stable for up to 3 months at –20 °C

Specification Test Results

About this item

 :
 :590.634 g/mol
Storage Temperature:Refrigerator
CAS Number:70967-90-7
Formula:C₂₇H₃₈N₆O₉
MDL Number:MFCD00077137