Order Entry
Germany
ContactUsLinkComponent
 
Analogue oven thermometer
  MOIN1455T
 :  MOINEAU
undefined
Analogue oven thermometer
  MOIN1455T
 :  MOINEAU

 

  • Description:
    Special oven thermometer, aluminium, 70×75 mm
  • Pk:
    1
  • Range:
    0...300 °C
  • Weight:
    40 g

 

 

Baking/cooking thermometer made from aluminium for use in ovens.