VWR® Maximum, Protection Covers
Supplier: VWR Collection
Certificates
About this item
Manufactured from fluid-impervious material.
- Full protection from fluids and light chemical splashes
- Lowest level of particle shedding
- Clean, low linting fabric
- Tunnelled elastic, along the bottom perimeter, ensures a secure fit
These covers provide barrier protection against fluids, biologicals, particulates and light chemical splashes.
Bacterial Filtration Efficiency: 99,9%
Level I per Helmke Drum test
Certifications: Covers are rigorously tested and manufactured in an ISO certified facility, under stringent process controls, to ensure that each item meets exacting quality standards, and performs to specification.
Lot-controlled and validated through independent lab testing.
Lot-controlled and validated through independent lab testing.