VWR® Maximum, Chemical Resistant Cover
Catalog # 129-1217
Supplier: VWR Collection
Specifications
- Colour:Yellow
- Description:Protection covers, non sterile
- W×D×H:1830×1530×1530 mm
- Pk:25
Specifications
About this item
These chemical resistant covers provide the highest level of protection available.
- Superior chemically repellent, as well as liquid and particle hold out
- Double-coated fabric protects against hazardous liquids and toxic chemicals
- Maximum durability and tear strength
These covers are designed to optimise barrier protection, and prevent penetration by an extensive list of chemicals.
Covers are rigorously tested and manufactured in an ISO certified facility, under stringent process controls, to ensure that each item meets exacting quality standards, and performs to specification.
Lot-controlled and validated through independent lab testing.