Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Reference materials, cloud point reference, CRM
Reference materials, cloud point reference, CRM
Reference materials, cloud point reference, CRM
HERZ01000-815-51
:
HERZOG WALTER
:
Reference materials, cloud point reference, CRM
HERZ01000-815-51
:
HERZOG WALTER
:
Pk:
1
Description:
Diesel fuel, −5 °C, 255 ml
:
ISO 3015; ASTM D 2500; ASTM D 5771; ASTM D 5772; DIN EN 23015 certified.