Industrial gloves, ActivArmr® 97-631
Supplier: Ansell
Certificates
About this item
These nylon/acrylic gloves with palm PVC coating provide mechanical and cold protection.
- PVC coating (palm dipped) stays flexible in cold temperatures
- Warme glove that helps to fight frostbite
- Industry leading combination of comfort, warmth, and dexterity
Typical applications include: Agriculture, cold storage, construction work, outdoor winter use, utilities.
Certifications: EN 388:2016 (2231B), EN 511 (12X); Cat. II
Food contact approved
Food contact approved
Specifications
- Catalog No:
- ANSE97-631/7
- ANSE97-631/9
- ANSE97-631/10
- ANSE97-631/11
- ANSE97-631/8
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/acrylic
- Nylon/acrylic
- Nylon/acrylic
- Nylon/acrylic
- Nylon/acrylic
- Finish:
- PVC coating on palm
- PVC coating on palm
- PVC coating on palm
- PVC coating on palm
- PVC coating on palm
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2231B), EN 511 (12X)
- EN 388:2016 (2231B), EN 511 (12X)
- EN 388:2016 (2231B), EN 511 (12X)
- EN 388:2016 (2231B), EN 511 (12X)
- EN 388:2016 (2231B), EN 511 (12X)
- Food contact approved:
- +
- +
- +
- +
- +