HPLC columns, YMC-Triart, C18 (ODS), 3 µm
Supplier: YMC
Certificates
About this item
YMC-Triart C18 HPLC columns have balanced hydrophobicity and silanol activity, which makes them for the ideal choice for the separation of acid, neutral, basic and chelating compounds. In addition to that these can also behave as a versatile stationary phase.
- Effective for analysing high polar compounds
- Effective for high sensitivity analysis using LC/MS
- High physical and chemical durability
These columns could be able to withstand at pH values of 1 - 12, at temperatures up to 70 °C and even with 100% aqueous eluents.
* high pressure rated HPLC column.