Standard hydrometers
Supplier: Alla France
Jump to:
About this item
These standard hydrometers strictly adhere to the principles of manufacture and specified calibration.
- Reading below meniscus
- Calibration temperature of 20 °C
- Reference mark on the stem shows any movement of the measurement scale
Certifications: NF, ISO, BS and DIN.
Delivery information: Supplied with certificate of conformity to ISO 649 (corresponding to BS718, DIN12791, NF B 35511).
22 Options Available Below
Product Details & Documents
These standard hydrometers strictly adhere to the principles of manufacture and specified calibration.
- Reading below meniscus
- Calibration temperature of 20 °C
- Reference mark on the stem shows any movement of the measurement scale
Certifications: NF, ISO, BS and DIN.
Delivery information: Supplied with certificate of conformity to ISO 649 (corresponding to BS718, DIN12791, NF B 35511).
Product Family Options
M50
Product Information
- ModelRangeDivisionLengthAvailabilityPrice
- ALLA3500MC06520BEUTraceable calibration certificateTraceable calibration certificateM500,650 - 0,700 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC07020BEUTraceable calibration certificateTraceable calibration certificateM500,700 - 0,750 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC07520BEUTraceable calibration certificateTraceable calibration certificateM500,750 - 0,800 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC08020BEUTraceable calibration certificateTraceable calibration certificateM500,800 - 0,850 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC08520BEUTraceable calibration certificateTraceable calibration certificateM500,850 - 0,900 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC09020BEUTraceable calibration certificateTraceable calibration certificateM500,900 - 0,950 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST447,00 €
- ALLA3500MC14520HEUTraceable calibration certificateTraceable calibration certificateM501,450 - 1,500 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST458,00 €
- ALLA3500MC14520BEUTraceable calibration certificateTraceable calibration certificateM501,450 - 1,500 g/ml0,001 g/ml270 mmLog in for availability1 * 1 ST458,00 €
- ALLA3500LC10020MEUTraceable calibration certificateTraceable calibration certificateL501,000 - 1,050 g/ml0,0005 g/ml335 mmLog in for availability1 * 1 ST477,00 €
M100
Product Information
- ModelRangeDivisionLengthAvailabilityPrice
- ALLA3100MC08020BEOOfficial calibration certificateOfficial calibration certificateM1000,800 - 0,900 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST1 200,00 €
- ALLA3100MC06020BEUTraceable calibration certificateTraceable calibration certificateM1000,600 - 0,700 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST451,00 €
- ALLA3100MC07020BEUTraceable calibration certificateTraceable calibration certificateM1000,700 - 0,800 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST447,00 €
- ALLA3100MC08020BEUTraceable calibration certificateTraceable calibration certificateM1000,800 - 0,900 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST447,00 €
- ALLA3100MC09020BEUTraceable calibration certificateTraceable calibration certificateM1000,900 - 1,000 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST447,00 €
- ALLA3100MC10020MEUTraceable calibration certificateTraceable calibration certificateM1001,000 - 1,100 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST445,00 €
- ALLA3100MC11020MEUTraceable calibration certificateTraceable calibration certificateM1001,100 - 1,200 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST445,00 €
- ALLA3100MC12020MEUTraceable calibration certificateTraceable calibration certificateM1001,200 - 1,300 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST445,00 €
- ALLA3100MC13020HEUTraceable calibration certificateTraceable calibration certificateM1001,300 - 1,400 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST451,00 €
- ALLA3100MC13020MEUTraceable calibration certificateTraceable calibration certificateM1001,300 - 1,400 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST451,00 €
- ALLA3100MC14020HEUTraceable calibration certificateTraceable calibration certificateM1001,400 - 1,500 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST451,00 €
- ALLA3100MC16020HEUTraceable calibration certificateTraceable calibration certificateM1001,600 - 1,700 g/ml0,002 g/ml250 mmLog in for availability1 * 1 ST460,00 €
LC50
Product Information
- ModelRangeDivisionLengthAvailabilityPrice
- ALLA3500LC08520BEUTraceable calibration certificateTraceable calibration certificateL500,850 - 0,900 g/ml0,0005 g/ml335 mmLog in for availability1 * 1 ST474,00 €