ContactUsLinkComponent
Organic reference standards, PCBs
Organic reference standards, PCBs
Catalog # PROOSL32110
CAS Number:  
Organic reference standards, PCBs
Catalog # PROOSL32110
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Standards:
    PCB standards
  • Description:
    Six key PCB isomers
Specifications
About this item
These standards contains six key PCB isomers such as 2,4,4'-trichlorobiphenyl; 2,2',5,5'-tetrachlorobiphenyl; 2,2',4,5,5'-pentachlorobipheny; 2,2',3,4,4',5'-hexachlorobiphenyl; 2,2',4,4',5,5'-hexachlorobiphenyl and 2,2',3,4,4',5,5'-heptachlorobiphenyl dissolved in iso-octane.