Order Entry
Germany
ContactUsLinkComponent
Streptomyces sp. N174 chitosanase (from Streptomyces sp. N174)
Streptomyces sp. N174 chitosanase (from Streptomyces sp. N174)
Catalog # 220477-10
CAS Number:  
Streptomyces sp. N174 chitosanase (from Streptomyces sp. N174)
Catalog # 220477-10
CAS Number:  

Specifications

  • Pk:
    10 EU
  • CAS No.:
    51570-20-8
  • Source:
    Streptomyces sp. N174
  • Species:
    Streptomyces sp. N174
  • Storage conditions:
    −20 °C
  • Form:
    Liquid
  • Enzyme name:
    Chitosanase
  • Enzyme activity:
    ≥25 units/mg protein
  • Enzyme synonyms:
    Chitosan N-acetylglucosaminohydrolase

Specifications

About this item

Chitosanase from Streptomyces species is a new class of enzymes with antifungal properties that efficiently catalyses the hydrolysis of acetylated chitosan.

  • Does not hydrolyse chitin or carboxylmethylcellulose
  • Activity increases with decreasing acetylation of chitosan

It acts by splitting the β1,4-glycosidic linkages of GlcNAc-GlcN and GlcNAc-GlcN. Suitable for the efficient preparation of protoplasts of Mucor and other Zygomycetes, allowing the genetic manipulation of these fungi.

Unit definition: One unit is defined as the amount of enzyme that will release 1,0 µmol reducing sugars or glucosamine from chitosan per min at 37 °C, pH 5,5.