Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Diastase (from fungi)
Diastase (from fungi)
Diastase (from fungi)
Catalog # 1.03604.0050
Supplier:
Merck
CAS Number:
SDS
Expand image
Diastase (from fungi)
Catalog # 1.03604.0050
Supplier:
Merck
CAS Number:
SDS
Specifications
Pk:
50 g
CAS No.:
9000-90-2
Source:
Fungi
Storage conditions:
2...8 °C
Form:
Brownish colour
Enzyme name:
Diastase
Solubility:
10 g/L
pH:
6,5 - 7,5
Specifications
About this item
Product Details
Documentation
Product Details
Documentation