Order Entry
Germany
ContactUsLinkComponent
Organic reference standards, canthaxanthine, Dr. Ehrenstorfer
Organic reference standards, canthaxanthine, Dr. Ehrenstorfer
  EHERCA10947000
 :  
Organic reference standards, canthaxanthine, Dr. Ehrenstorfer
  EHERCA10947000
 :  

 

  • Pk:
    0,15 g
  • Standards:
    Reference standards
  • CAS No.:
    514-78-3
  • Description:
    Canthaxanthine
  • Formula:
    C₄₀H₅₂O₂
  • Description:
    Canthaxanthine