Order Entry
Germany
ContactUsLinkComponent
Xenon ≥99.99%, Messer® CANGas
Xenon ≥99.99%, Messer® CANGas
Catalog # SIAL769118-1L
Supplier:  Merck
CAS Number:  
Xenon ≥99.99%, Messer® CANGas
Catalog # SIAL769118-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • CAS Number:
    7440-63-3
  • Empirical Formula (Hill Notation):
    Xe
  • Molecular Weight:
    131,29
  • EC Number:
    231-172-7
  • MDL number:
    MFCD00036280
  • PubChem Substance ID:
    329767434
  • Filling mass:
    71 g
  • Filling volume:
    12 l
  • InChI Key:
    FHNFHKCVQCLJFQ-UHFFFAOYSA-N
  • Assay:
    99,99%

Specification Test Results

About this item

 :
 :131.29 g/mol
UN:2036
ADR:2,
CAS Number:7440-63-3
Formula:Xe
MDL Number:MFCD00036280