Methyl oleate 99% analytical standard for GC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-62-9
:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578
:296.49 g/mol
Boiling Pt:186 °C (9 mmHg)
Melting Pt:–20 °C
Density:0.87 g/cm³
Flash Pt:180 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4530 (20°C, 589 nm)
- GC:≥98.5 %