Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Certificates
undefined
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed

Specification Test Results

  • Appearance:
    Off-white to light yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Assay:
    98 to 98.8 mol % (degree of hydrolysis)
  • Viscosity:
    3.5 to 4.5 cP (4 % in water)
  • Ash:
    ≤1.2 % (as Na2O)
  • pH:
    5.0 to 7.0 (4 % in water)
  • Volatile matter:
    ≤5 %

Specification Test Results