Chemical resistant overalls, Tychem® 6000 F FaceSeal, model TF 611 T
Supplier: DuPont
Certificates
About this item
These garments are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Hooded overall with rubber seal on hood for tight fit around respirator
- Back entry with zipper, double zipper flap with snaps and hook & loop closure
- Attached undergloves and dissipative socks with boot flap
- Elasticated waist for an optimal fit
- Stitched and over-taped seams with barrier tape for protection and strength
- Anti-static treatment (EN 1149-5) on inside
Tychem® F FaceSeal can be used for a broad range of applications from industrial manufacturing and cleaning, emergency response, chemical spill clean-up to military and petrochemical applications.
Weight: 120 g/m²
Colour: Grey
Certifications: Type 3B/4B/5B/6B, EN 1149-5, EN 1073-2, EN 14126, Cat. III; (EU) 2016/425
Packaging: 1 per box.
Specifications
- Catalog No:
- DUPOTF0611TGYUG/4X
- DUPOTF0611TGYUG/XL
- DUPOTF0611TGYUG/L
- DUPOTF0611TGYUG/M
- DUPOTF0611TGYUG/S
- DUPOTF0611TGYUG/2X
- DUPOTF0611TGYUG/3X
- DUPOTF0611TGYUG/5X
- Model:
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- 6000 F FaceSeal, model TF 611 T
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Weight:
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²