Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Graphene oxide
Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Supplier:
Merck
About this item
:
Formula:
CxOyHz