Organic reference standards, α-zearalenol (NMIA P1795)
Catalog # PROONMIAP1795
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:5 mg
- Standards:Reference standard
- CAS No.:36455-72-8
- Description:α-Zearalenol
- Formula:C₁₈H₂₄O₅
- Description:α-Zearalenol
Specifications
About this item
Purity estimate obtained by subtraction from 100% of total impurities by GC-FID, thermogravimetric analysis, Karl Fischer analysis and 1H NMR is 99,1 ± 1,4% mass fraction, (95% coverage interval). Supporting evidence is provided by elemental microanalysis.
- White crystalline powder
- Moisture content ≤0,5% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,2% mass fraction
- CAS no.: 36455-72-8
Synonyms: (3S,7R,11E)-3,4,5,6,7,8,9,10-octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one.