Order Entry
Germany
ContactUsLinkComponent
Organic reference standards, fipronil sulphone (NMIA P1731)
Organic reference standards, fipronil sulphone (NMIA P1731)
  PROONMIAP1731
 :  
Organic reference standards, fipronil sulphone (NMIA P1731)
  PROONMIAP1731
 :  

 

  • Pk:
    25 mg
  • Standards:
    Reference standard
  • CAS No.:
    424-610-5
  • Description:
    Fipronil sulphone
  • Formula:
    C₁₂H₄Cl₂F₆N₄O₂S
  • Description:
    Fipronil sulphone

 

 

Purity estimate obtained from a combination of traditional analytical techniques is 99,3 ± 0,6% mass fraction, (95% coverage interval). The purity estimate by traditional analytical techniques was obtained by subtraction from 100% of total impurities by GC-FID, and Karl Fischer analysis. Supporting evidence is provided by elemental microanalysis and 1H NMR analysis.

  • White solid
  • Moisture content ≤0,2% mass fraction
  • Volatile residue ≤0,1% mass fraction
  • Non volatile residue ≤0,2% mass fraction

Synonym: 5-amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-4-((trifluoromethyl)sulphonyl)pyrazole-3-carbonitrile.