ContactUsLinkComponent
ATP assay kit
ATP assay kit
Catalog # ABNOKA1661
Supplier:  Abnova
CAS Number:  
undefined
ATP assay kit
Catalog # ABNOKA1661
Supplier:  Abnova
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    ATP Assay Kit
  • Tests per kit:
    100
Specifications
About this item
ATP Assay Kit is for the rapid bioluminescent determination of ATP in biological samples.
Delivery: Kit contains assay buffer (10 ml), substrate (120 µl), ATP enzyme (120 µl), ATP standard (120 µl).