Order Entry
Germany
Orders LinkContactUsLinkComponent
Exemestane, Reference Standards

Exemestane, Reference Standards

Supplier:   USP
Exemestane, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1269050.
  • 1269094.
  • Description:
  • Exemestane
  • Exemestane System Suitability Mixture
  • CAS No.:
  • 107868-30-4
  • Formula:
  • C₂₀H₂₄O₂

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1269050.
    Exemestane
    Exemestane
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 404,00 €
    404,00 €
    1 * 150 mg
       404,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269050.
    Description
    Exemestane
    CAS No.
    107868-30-4
    Formula
    C₂₀H₂₄O₂
  • 1269094.
    Exemestane System Suitability Mixture
    Exemestane System Suitability Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 360,00 €
    1 360,00 €
    1 * 25 mg
       1 360,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269094.
    Description
    Exemestane System Suitability Mixture
    CAS No.
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.