Order Entry
Germany
ContactUsLinkComponent
 
Solvent blue 35, violet crystalline powder, Sigma-Aldrich®
 
undefined
Solvent blue 35, violet crystalline powder, Sigma-Aldrich®
 

 

  • Composition :
    Dye content, 98%
  • Absorption :
    λmax 652 nm
  • Appearance (color) :
    Maroon to violet or purple
  • Appearance (form) :
    Powder or crystalline powder
  • Infrared spectrum :
    Conforms to structure
  • Wavelength, c = 0.01g/l in chloroform:
    648 - 652 nm
  • Extinction coefficient:
    ≥16000
  • Wavelength:
    600 - 604 nm
  • Carbon :
    73,5 - 77,3%
  • Nitrogen :
    7,7 - 8,3%
  • Solubility (turbidity), c = 4 mg/ml in chloroform, clear to opaque:
    Conforms
  • Solubility (color) :
    Blue