Cryoboxes, Plastic Coated, 148×148 mm
Supplier: RATIOLAB
Certificates
About this item
Cryoboxes, cryo resistant cardboard.
- Various combinations possible with different grid inserts
- Height stated is the interior height
- Supplied with lid
- Cryo boxes can be used for larger vessels when grid is removed
- Temperature resistant down to – 80 °C
For storage of cryovials at low temperatures.