Organic reference standard, Hyoscine hydrobromide, Ph.Eur., RS
Catalog # EDQMH1500000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:51-56-9
- Description:Hyoscine hydrobromide
- Formula:C₁₆H₂₂BrNO₃
- Description:Hyoscine hydrobromide
Specifications
About this item
- In Monographs 0106 and 0500
- Solubility in water: 33,3 to 100 g/l