Wipes rolls, WYPALL* L30 ULTRA+
Supplier: KIMBERLY CLARK
Certificates
About this item
These single use wipes made from three ply AIRFLEX* material are ideal for superior cleaning and heavy duty wiping tasks such as soaking up industrial spills, cleaning parts and components.
- Strongest, thickest and most absorbent WYPALL L wipe
- Stronger than conventional three ply wipes
- Available as a white or blue perforated large roll
- Can be used with a dispensing system to help control use and reduce wastage
For use with dispensers 115-0045, 115-1533 and 115-2038.
Related Information
KIMB7331_TDS_EN KIMB7426_TDS_EN
Datasheet
Kimberly-Clark Kimtech™ Lab Gloves brochure