ContactUsLinkComponent
Human serum, colorectal cancer (CRC)
Human serum, colorectal cancer (CRC)
Catalog # PB007C-1-CRC
Human serum, colorectal cancer (CRC)
Catalog # PB007C-1-CRC
Specifications
  • Serum format:
    Cryopreserved
  • Description:
    Serum, colorectal cancer (CRC)
  • Pk:
    2 mL
Specifications
About this item
Human serum used for researching the causes, diagnosis, supportive care and treatment of colorectal cancer (CRC). Collected from IRB-consented donors diagnosed with CRC.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.