Order Entry
Germany
Orders LinkContactUsLinkComponent
Lisinopril, Reference Standards

Lisinopril, Reference Standards

Supplier:   USP
Lisinopril, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1368609.
  • 1368610.
  • Description:
  • Lisinopril
  • Lisinopril Related Compound A
  • Synonym(s):
  • (S)-2-{(3S,8aS)-3-(4-aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-α]pyrazin-2(1H)-yl}-4-phenylbutanoic acid
  • CAS No.:
  • 83915-83-7
  • 328385-86-0
  • Formula:
  • C₂₁H₃₅N₃O₇
  • C₂₁H₂₉N₃O₄

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1368609.
    Lisinopril
    Lisinopril
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 385,00 €
    385,00 €
    1 * 300 mg
       385,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1368609.
    Description
    Lisinopril
    Synonym(s)
    CAS No.
    83915-83-7
    Formula
    C₂₁H₃₅N₃O₇
  • 1368610.
    Lisinopril Related Compound A
    Lisinopril Related Compound A
    USP reference standard
    Vial
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 1 410,00 €
    1 410,00 €
    1 * 250 mg
       1 410,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1368610.
    Description
    Lisinopril Related Compound A
    Synonym(s)
    (S)-2-{(3S,8aS)-3-(4-aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-α]pyrazin-2(1H)-yl}-4-phenylbutanoic acid
    CAS No.
    328385-86-0
    Formula
    C₂₁H₂₉N₃O₄
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.