Labels for safety padlocks
Supplier: Brady
Certificates
About this item
These vinyl adhesive labels help send a clear message to workers when a machine is down for service or maintenance.
- Available in three languages (English, French, Spanish)
- For use with safety padlocks
Colour: Black and red on white
Thickness: 0,124 mm