ContactUsLinkComponent
Elemental analysis standards, plastic clay, NIST-98B
Elemental analysis standards, plastic clay, NIST-98B
Catalog # PROONIST-98B
CAS Number:  
Elemental analysis standards, plastic clay, NIST-98B
Catalog # PROONIST-98B
CAS Number:  
Specifications
  • Pk:
    60 g
  • Description:
    NIST-98B Plastic clay - constituent elements
Specifications
About this item
This standards reference material is intended to use in the determination of constituent elements in clay and material of similar matrix. These powdered clay was air-dried, ball-milled and blended to ensure homogeneity.
Certifications: Traceable to NIST and SRM.