Haloperidol, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1303002.
- 1303013.
- 1303024.
- 1303057.
- Description:
- Haloperidol
- Haloperidol Related Compound A
- Haloperidol Related Compound B
- Haloperidol Decanoate
- Synonym(s):
- 4,4'-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]-butyrophenone
- 4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one
- CAS No.:
- 52-86-8
- 67987-08-0
- 1391052-53-1
- 74050-97-8
- Formula:
- C₂₁H₂₃ClFNO₂
- C₃₂H₃₆Cl₂N₂O₃
- C₂₁H₂₃ClFNO₂
- C₃₁H₄₁ClFNO₃
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1303002.HaloperidolSpecifications:
More
Specifications
Cat. No.1303002.DescriptionHaloperidolSynonym(s)CAS No.52-86-8FormulaC₂₁H₂₃ClFNO₂UN No.2811 - 1303013.Haloperidol Related Compound AHaloperidol Related Compound AUSP reference standardVial10 mgLog in for availability1 * 10 mg1 230,00 €Specifications:
More
Specifications
Cat. No.1303013.DescriptionHaloperidol Related Compound ASynonym(s)4,4'-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]-butyrophenoneCAS No.67987-08-0FormulaC₃₂H₃₆Cl₂N₂O₃UN No. - 1303024.Haloperidol Related Compound BHaloperidol Related Compound BUSP reference standardVial10 mgLog in for availability1 * 10 mg1 200,00 €Specifications:
More
Specifications
Cat. No.1303024.DescriptionHaloperidol Related Compound BSynonym(s)4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-oneCAS No.1391052-53-1FormulaC₂₁H₂₃ClFNO₂UN No. - 1303057.Haloperidol DecanoateSpecifications:
More
Specifications
Cat. No.1303057.DescriptionHaloperidol DecanoateSynonym(s)CAS No.74050-97-8FormulaC₃₁H₄₁ClFNO₃UN No.



