Flurbiprofen, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1285750.
- 1285760.
- 1285807.
- Description:
- Flurbiprofen
- Flurbiprofen Related Compound A
- Flurbiprofen Sodium
- Synonym(s):
- 2-(4-Biphenylyl)propionic Acid
- CAS No.:
- 5104-49-4
- 6341-72-6
- 56767-76-1
- Formula:
- C₁₅H₁₄O₂
- C₁₅H₁₂FNaO₂
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1285750.FlurbiprofenSpecifications:
More
Specifications
Cat. No.1285750.DescriptionFlurbiprofenSynonym(s)CAS No.5104-49-4FormulaUN No.2811 - 1285760.Flurbiprofen Related Compound AFlurbiprofen Related Compound AUSP reference standardVial100 mgLog in for availability1 * 100 mg1 250,00 €Specifications:
More
Specifications
Cat. No.1285760.DescriptionFlurbiprofen Related Compound ASynonym(s)2-(4-Biphenylyl)propionic AcidCAS No.6341-72-6FormulaC₁₅H₁₄O₂UN No. - 1285807.Flurbiprofen SodiumSpecifications:
More
Specifications
Cat. No.1285807.DescriptionFlurbiprofen SodiumSynonym(s)CAS No.56767-76-1FormulaC₁₅H₁₂FNaO₂UN No.



