Order Entry
Germany
ContactUsLinkComponent
H. influenzae recombinant NAD Nucleotidase (from E. coli)
H. influenzae recombinant NAD Nucleotidase (from E. coli)
Catalog # PRSI90-368
Supplier:  ProSci Inc.
CAS Number:  
H. influenzae recombinant NAD Nucleotidase (from E. coli)
Catalog # PRSI90-368
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    H. influenzae
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAD Nucleotidase
  • Enzyme activity:
    With AMP: 856.4 nmol of Ade/min/mg of protein
    With NMN: 110,3 nmol of NR/min/mg of protein
    With NAD: 5.4 nmol of Ade/min per mg of protein | 5.3 nmol of NR/min/mg of protein
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    Full-length NAD 5’-nucleotidase without the signal peptide corresponding to the first 26 aa
  • Formulation:
    Liquid, In 5 mM KH₂PO₄, pH 6.8 containing 100 mM TRIS-HCl (pH 8.5) and 10 mM MgCl₂.

Specifications

About this item

NadN (NAD nucleotidase) is a periplasmic enzyme from Haemophilus influenzae, a major pathogen of the respiratory tract in humans that has developed the capability to exploit host NAD(P) for its nicotinamide dinucleotide requirement. NadN plays a central role by degrading NAD into adenosine and NR (nicotinamide riboside), NMN to nicotinamide riboside and AMP to adenosine. It shows a broad substrate specificity, recognising either mono- or di-nucleotide nicotinamides and different adenosine phosphates with a maximal activity on 5'-adenosine monophosphate. It can be used as an economically alternative to produce NR (nicotinamide riboside). Catalytic activity: NAD=AMP + NMN=Ade+NR+PPi | AMP=Ade+Pi | NMN=NR+Pi.

Fusion-Tag: Tag Free

This recombinant proteins is for research use only.