Mouse recombinant carboxypeptidase A2
Catalog # PRSI92-419
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Carboxypeptidase A2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:46.2 kDa
- Sequence:Gln17-Tyr417
- Formulation:Lyophilized from a 0,2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
Mouse carboxypeptidase A2(CPA2) is a secreted pancreatic procarboxy -peptidase which belongs to the peptidase M14 family. CPA2 consists of a signal peptide, a pro region and a mature chain. It can be activated after cleavage of the pro peptide. CPA2 cleaves the C-terminal amide or ester bond of peptides that have a free C-terminal carboxyl group. The hydrolytic action of CPA2 was identified with a preference towards long substrates with aromatic amino acids in their C-terminal end, particularly tryptophan.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.