Mouse recombinant darcin (from E. coli)
Catalog # PRSI90-264
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:Tag Free
- Storage conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Protein synonyms:Mup20|Major Urinary Protein 24|Mouse Urinary Protein 20
- Protein/peptide name:Darcin
- Purity:>95% (SDS-PAGE)
- Sequence:20-181
- Formulation:Lyophilized, Contains PBS.
Specifications
About this item
Darcin (also called Mup20) belongs to the family of major urinary proteins (MUPs), a subfamily of proteins found in abundance in the urine. Like all MUPs, Darcin is a member of the lipocalin family of ligand-binding proteins, and preferentially binds the volatile pheromone (S)-2-sec-butyl-4,5-dihydrothiazole (SBT). Darcin is present in the urine of male and acts as a sex pheromone that elicits female sexual attraction to a male's urine scent. Recombinant Darcin shows similar activity compared to natural Darcin.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.