Human Recombinant CD137 (from HEK293 cells)
PRSI90-252
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Protein synonyms:TNFRSF9|4-1BB
- Protein/peptide name:CD137
- Purity:>95% (SDS-PAGE)
- Sequence:1-186
- Formulation:Lyophilized, Contains PBS.
CD137 (4-1BB) is a member of the tumor necrosis factor (TNF) receptor family. CD137 is expressed by activated T cells, dendritic cells, NK cells, granulocytes and cells of blood vessel walls at sites of inflammation. CD137 has costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. CD137 was shown to enhance immune activity to eliminate tumours in mice.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.