Order Entry
Germany
ContactUsLinkComponent
Human recombinant OX40 (from cells)
Human recombinant OX40 (from cells)
Catalog # PRSI92-553
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant OX40 (from cells)
Catalog # PRSI92-553
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    OX40|Tumor necrosis factor receptor superfamily member 4|Txgp1|REF: C1130|TNFRSF4|CD134
  • Protein/peptide name:
    OX40
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    46.8 kD
  • Sequence:
    Leu29-Ala216
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

OX40, also termed CD134 and TNFRSF4, is a T cell co-stimulatory molecule of the TNF receptor superfamily which plays a key role in the survival and homeostasis of effector and memory T cells. OX40 is expressed on CD4+ and CD8+ T cells upon engagement of the TCR by antigen presenting cells along with co-stimulation by CD40-CD40 Ligand and CD28-B7. The interaction between OX40 and OX40 ligand (OX40 L) will occur when activated T cells bind to professional antigen-presenting cells (APCs). The T-cell functions, including cytokine production, expansion, and survival, are then enhanced by the OX40 costimulatory signals. OX40 signals are critical for controllling the function and differentiation of Foxp3+ regulatory T cells. OX40-OX40L interaction regulates T-cell tolerance, peripheral T-cell homeostasis, and T-cell-mediated inflammatory diseases.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.