Order Entry
Germany
ContactUsLinkComponent
 
Human recombinant IL-12 R beta1 (from Cells)
Catalog # PRSI92-544
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-12 R beta1 (from Cells)
Catalog # PRSI92-544
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-12R subunit beta-1|IL-12R-beta-1|CD212|IL12RB|CD212 antigen|IL-12R-BETA1|IL-12 receptor beta component|IL12RB1|interleukin-12 receptor beta-1 chain|IL-12RB1|interleukin-12 receptor subunit beta-1|IL12R|IL-12 receptor subunit beta-1
  • Protein/peptide name:
    IL-12 R beta1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    84.2 kD
  • Sequence:
    Cys24-Glu540
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Interleukin12 receptor subunit beta 1 (IL12RB1) is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. IL12RB1 can spontaneously form homodimers and -oligomers, which are able to bind IL12 with only low affinity. IL12 high affinity receptor complex is composed of two subunits designated IL12RB1 and IL12RB2. While IL12RB1 interacts with the IL-12p40 subunit, IL-12p35 is mainly connecting with IL12RB2. This receptor chain is also responsible for transmitting the IL12 signal into the cell. IL12RB1, to the contrary, is also part of the IL23R, where it interacts with the p40 subunit of IL23. IL12RB1 is expressed in activated T cells, NK cells and B cells.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.