Order Entry
Germany
ContactUsLinkComponent
Human recombinant IFN gamma R1 (from cells)
Human recombinant IFN gamma R1 (from cells)
Catalog # PRSI92-375
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IFN gamma R1 (from cells)
Catalog # PRSI92-375
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Interferon gamma receptor 1|IFNGR1|CD119|CDw119
  • Protein/peptide name:
    IFN gamma R1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.7 kD
  • Sequence:
    Glu18-Ser245
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Interferon gamma receptor 1(IFNGR1) encoded by the IFNGR1 gene, is a single-pass type 1 membrane protein which belongs to the type II cytokine receptor family. IFNGR1 is phosphorylated at Ser/Thr residues after translation. IFNGR1 is a receptor for interferon gamma, two receptors bind one interferon gama dimer. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.