Human recombinant Endoplasmic Reticulum Resident 44 (from cells)
Catalog # PRSI91-923
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:KIAA0573|Thioredoxin domain-containing protein 4|ER protein 44|ER resident protein 44|ERp44TXNDC4|TXNDC4
- Protein/peptide name:Endoplasmic Reticulum Resident 44
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:44.2 kD
- Sequence:Glu30-Asp402
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris HCl,10%glycerol,PH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Endoplasmic reticulum resident protein 44 (TXNDC4) is is a 406 amino acid protein that contains one thioredoxin domain. TXNDC4 mediates thiol-dependent retention in the early secretory pathway and forms mixed disulfides with substrate proteins through its conserved CRFS motif.It can inhibit the calcium channel activity of ITPR1.It may have a role in the control of oxidative protein folding in the endoplasmic reticulum.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag