Human recombinant Mucin 15 (from cells)
Catalog # PRSI91-727
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:MUC15|Mucin-15|MUC-15
- Protein/peptide name:Mucin 15
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:24.14 kD
- Sequence:Lys24-Thr236
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Mucin-15 is a single-pass type I membrane protein member of the Mucin family. Mucins are a family of high molecular weight, heavily glycosylated proteins (glycoconjugates) produced by epithelial tissues in most metazoans. A key characteristic of Mucins is their ability to form gels. Therefore they are a key component in most gel-like secretions, serving functions from lubrication to cell signalling to forming chemical barriers. Mucin-15 is expressed in many tissues. Mucin-15 is highly glycosylated (N- and O-linked carbohydrates). Mucin-15 may play a role in the cell adhesion to the extracellular matrix.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag