Order Entry
Germany
ContactUsLinkComponent
Human recombinant Folate Receptor alpha (from cells)
Human recombinant Folate Receptor alpha (from cells)
Catalog # PRSI91-626
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Folate Receptor alpha (from cells)
Catalog # PRSI91-626
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ovarian tumor-associated antigen MOv18|FOLR1|Folate receptor alpha|Adult folate-binding protein|Folate receptor 1|FBP|FR-alpha|Folate receptor
  • Protein/peptide name:
    Folate Receptor alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.7 kD
  • Sequence:
    Arg25-Ser234
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Folate receptor alpha(FOLR) belongs to the folate receptor family, and is primarily expressed in tissues of epithelial origin. It is also expressed in kidney, lung and cerebellum. The secreted form is derived from the membrane-bound form either by cleavage of the GPI anchor, or/and by proteolysis catalyzed by a metalloprotease. FOLR1 binds to folate and reduced folic acid derivatives and mediates delivery of 5-methyltetrahydrofolate and folate analogs into the interior of cells. It has high affinity for folate and folic acid analogs at neutral pH. Exposure to slightly acidic pH after receptor endocytosis triggers a conformation change that strongly reduces its affinity for folates and mediates their release. It is required for normal embryonic development and normal cell proliferation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag