Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:SynCAM1|Tumor suppressor in lung cancer 1|Nectin-like protein 2|Immunoglobulin superfamily member 4|TSLC-1|Cell adhesion molecule 1|SynCAM|NECL-2|Spermatogenic immunoglobulin superfamily|IGSF4A|IgSF4|Synaptic cell adhesion molecule|SgIgSF
- Protein/peptide name:CADM1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:65.3 kD
- Sequence:Gln48-His388
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Cell adhesion molecule 1(CADM1) is a single-pass type I membrane protein and belongs to the nectin family. It contains 2 Ig-like C2-type (immunoglobulin-like) domains and 1 Ig-like V-type (immunoglobulin-like) domain. CADM1 acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. CADM1 may contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, it may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. The protein acts as a synaptic cell adhesion molecule and plays a role in the formation of dendritic spines and in synapse assembly. It may be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. CADM1 may play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag