Mouse recombinant MMP-12
Catalog # PRSI91-577
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Mmp12|Macrophage metalloelastase|Mmel|MMP-12|Matrix metalloproteinase-12|MME
- Protein/peptide name:MMP-12
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:53 kD
- Sequence:Ala29-Cys473
- Formulation:Supplied as a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Matrix metalloproteinase-12(MMP12) is a secreted protein.It contains 4 hemopexin repeats and belongs to the peptidase M10A family. MMP12 may be involved in tissue injury and remodeling and have significant elastolytic activity. It can accept large and small amino acids at the P1' site, but has a preference for leucine. Aromatic or hydrophobic residues are preferred at the P1 site, with small hydrophobic residues (preferably alanine) occupying P3.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag