Human recombinant Cystatin S (from HEK293 cells)
Catalog # PRSI96-208
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:Cystatin-S5|Cystatin-S|CST4|Cystatin-4|Cystatin-SA-III
- Protein/peptide name:Cystatin S
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:15 kDa
- Formulation:Lyophilized, 25 mM MES, 150 mM NaCl, pH 6.0
- Tested applications:Western Blot
Specifications
About this item
Cystatin-4 (CST4) is also known as Cystatin-S, Salivary acidic protein 1, which is a secreted protein, which belongs to the cystatin family. Cystatin-S is expressed in submandibular and sublingual saliva but not in parotid saliva (at protein level) and is also expressed in saliva, tears, urine and seminal fluid. Cystatin-4 / Cystatin S strongly inhibits papain and ficin, partially inhibits stem bromelain and bovine cathepsin C, but does not inhibit porcine cathepsin B or clostripain. Papain is inhibited non-competitively.