Order Entry
Germany
ContactUsLinkComponent
Human recombinant CD36 (from HEK293 cells)
Human recombinant CD36 (from HEK293 cells)
Catalog # PRSI96-121
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant CD36 (from HEK293 cells)
Catalog # PRSI96-121
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized rhCD36 at 2μg/mL (100μL/well) can bind rhTSP2/His with a linear range of 0. 01-1μg/mL.
  • Protein synonyms:
    GP4|GP3B|FAT|GPIV|gpIV|SCARB3|CHDS7|glycoprotein IIIb|CD36|glycoprotein IV|BDPLT10|gpIIIb|PASIV|Platelet Glycoprotein 4
  • Protein/peptide name:
    CD36
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    47.5 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

CD36 (Cluster of Differentiation 36) is also known as platelet membrane glycoprotein IV (GPIV), fatty acid translocase (FAT), thrombospondin receptor, collagen receptor, and scavenger receptor class B, member 3 (SRB3), is a member of the class B scavenger receptor family of cell surface proteins. The human CD36 gene encodes a single chain 472 amino acid residue protein containing both an N- and a C-terminal cytoplasmic tail and an extracellular loop.CD36 is found on platelets, erythrocytes, monocytes, differentiated adipocytes, mammary epithelial cells, spleen cells and some skin microdermal endothelial cells. CD36 is a multiligand pattern recognition receptor that interacts with a large number of structurally dissimilar ligands, including long chain fatty acid (LCFA), advanced glycation end products (AGE), thrombospondin-1, oxidized low-density lipoproteins (oxLDLs), high density lipoprotein (HDL), phosphatidylserine, apoptotic cells, beta-amyloid fibrils (fAβ), collagens I and IV, and Plasmodium falciparum infected erythrocytes. CD36 is required for the anti-angiogenic effects of thrombospondin1 In the corneal neovascularisation assay. On binding a ligand the protein and ligand are internalized. This internalisation is independent of macropinocytosis and occurs by an actin dependent mechanism requiring the activation Src-family kinases, JNK and Rho-family GTPases. CD36 ligands have also been shown to promote sterile inflammation through assembly of a Toll-like receptor 4 and 6 heterodimer.