Order Entry
Germany
ContactUsLinkComponent
Human SCRN2 peptide (18 aa near C-terminus)
Human SCRN2 peptide (18 aa near C-terminus)
Catalog # PRSI5851P
Supplier:  ProSci Inc.
CAS Number:  
Human SCRN2 peptide (18 aa near C-terminus)
Catalog # PRSI5851P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa near C-terminus
  • Protein/peptide name:
    SCRN2 Peptide (18 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

SCRN2 peptide is used for blocking the activity of SCRN2 antibody.