Straining baskets, SP Bel-Art
Perforated baskets made from polyethylene plastic allow rapid filling and draining, and are handy for batch rinsing and transporting labware as well as straining or filtering large particles.
- Chemically resistant; withstand acids and alkalis
- Service temperature up to 82 °C
- With 2 mm perforations
- Strap handle rises 100 mm above the rim
Perforated baskets made from polyethylene plastic allow rapid filling and draining, and are handy for batch rinsing and transporting labware as well as straining or filtering large particles.
- Chemically resistant; withstand acids and alkalis
- Service temperature up to 82 °C
- With 2 mm perforations
- Strap handle rises 100 mm above the rim
Product Family Options
- DescriptionØ×H