Supelco® Drug and metabolite standards, O-desmethyltramadol hydrochloride, LoGiCal®
PROOLGCFOR0007.06
:
- Pk:10 mg
- Standards:Reference standard
- CAS No.:148262-77-5
- Description:O-Desmethyltramadol hydrochloride
- Formula:C₁₅H₂₄ClNO₂
- Description:O-Desmethyltramadol hydrochloride