Anti-CXCL9 Mouse Monoclonal Antibody [clone: 1.1_2D3-2H3]
PRSIXP-5721-M
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) ligand 9
- Antigen symbol:CXCL9
- Clonality:Monoclonal
- Clone:1.1_2D3-2H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:4283
- Antigen synonyms:SCYB9|HuMIG|crg-10|C-X-C motif chemokine 9|Gamma-interferon-induced monokine|MIG|Humig|CMK|monokine induced by interferon-gamma
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in Balb/c X ICR F1 mice using recombinant hMIG as the immunizing antigen.
- Purification:This IgG1K antibody was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
MIG, a member of the alpha-chemokine family (CXC) of cytokines, is produced by stimulated monocytes, macrophages and endothelial cells. It signals through the CXCR3 receptor.
Type: Primary
Antigen: CXCL9
Clonality: Monoclonal
Clone: 1.1_2D3-2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: