Anti-STAT5A Rabbit Polyclonal Antibody
Catalog # PRSI79-126
Supplier: ProSci Inc.
Specifications
- Antigen name:Signal Transducer And Activator Of Transcription 5A
- Antigen symbol:STAT5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:STAT5|MGF
- Concentration:1 mg/mL
- Immunogen:STAT5A (Phospho-Ser780) antibody was raised against a peptide sequence around phosphorylation site of serine 780 (R-L-S (p) -P-P) derived from Human STAT5A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Carries out a dual function: signal transduction and activation of transcription. Binds to the GAS element and activates PRL-induced transcription.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat